missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzothiazole, 97%, Thermo Scientific™
£12.90 - £48.20
Chemical Identifiers
CAS | 149-30-4 |
---|---|
Molecular Formula | C7H5NS2 |
Molecular Weight (g/mol) | 167.24 |
MDL Number | MFCD00005781 |
InChI Key | YXIWHUQXZSMYRE-UHFFFAOYSA-N |
Synonym | 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax |
PubChem CID | 697993 |
ChEBI | CHEBI:34292 |
IUPAC Name | 3H-1,3-benzothiazole-2-thione |
SMILES | C1=CC=C2C(=C1)NC(=S)S2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10754933
|
Thermo Scientific Acros
332050050 |
5g | Glass Bottle |
£12.90
5g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10275921
|
Thermo Scientific Acros
332050250 |
25g | Plastic bottle |
£14.50
25g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10526271
|
Thermo Scientific Acros
332051000 |
100g | Plastic bottle |
£16.50
100g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10380114
|
Thermo Scientific Acros
332050010 |
1kg | Plastic bottle |
£48.20
1kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
IndicatorChemical Identifiers
149-30-4 | |
167.24 | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
697993 | |
3H-1,3-benzothiazole-2-thione |
C7H5NS2 | |
MFCD00005781 | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
CHEBI:34292 | |
C1=CC=C2C(=C1)NC(=S)S2 |
Specifications
2-Mercaptobenzothiazole | |
149-30-4 | |
100.0 | |
172.0°C to 182.0°C | |
97% | |
C7H5NS2 | |
27, II, 233 | |
15, 5935 | |
5g | |
YXIWHUQXZSMYRE-UHFFFAOYSA-N | |
3H-1,3-benzothiazole-2-thione | |
697993 | |
167.24 | |
97% |
98% | |
97.5 | |
Yellow | |
243°C | |
Authentic | |
MFCD00005781 | |
Glass Bottle | |
2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax | |
yellow solution | |
C1=CC=C2C(=C1)NC(=S)S2 | |
167.24 | |
CHEBI:34292 | |
Powder |
RUO – Research Use Only