missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, 97%

Product Code. 11498026
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2500 g
Unit Size:
100g
10g
2500g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498026 100 g 100g
11379589 10 g 10g
11408036 500 g 500g
11418036 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11498026 Supplier Thermo Scientific Alfa Aesar Supplier No. A14086.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-30-4 | C7H5NS2 | 167.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.

Solubility
Soluble in ethanol, diethyl ether, benzene, carbon tetrachloride, alkali and alkaline solutions. Slightly soluble in acetone, naphtha and glacial acetic acid. Insoluble in water and gasoline.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.244
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

Melting Point 178°C to 182°C
Density 1.42
Flash Point 243°C (469°F)
Quantity 100 g
UN Number UN3077
Beilstein 119484
Sensitivity Air sensitive
Merck Index 14,5868
Solubility Information Soluble in ethanol,diethyl ether,benzene,carbon tetrachloride,alkali and alkaline solutions; Slightly soluble in acetone,naphtha and glacial acetic acid; Insoluble in water and gasoline.
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Mercaptobenzothiazole
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Mercaptobenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.