missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzoic Acid, TRC

Product Code. 30468957
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30468957 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30468957 Supplier Toronto Research Chemicals Supplier No. TRCM25170010G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Mercaptobenzoic Acid
CAS 147-93-3
Molecular Formula C7 H6 O2 S
Quantity 10 g
Synonym 2-Mercaptobenzoic acid (ACI), Benzoic acid, o-mercapto- (8CI), 2-Carboxybenzenethiol, 2-Carboxythiophenol, 2-Sulfanylbenzoic acid, 2-Thiosalicylic acid, NSC 2184, NSC 660640, o-Carboxythiophenol, o-Mercaptobenzoic acid, o-Sulfhydrylbenzoic acid, o-Thiosalicylic acid, Salicylic acid, 2-thio-, Thiophenol-2-carboxylic acid, Thiosalicylic acid
InChI Formula InChI=1S/C7H6O2S/c8-7(9)5-3-1-2-4-6(5)10/h1-4,10H,(H,8,9)
SMILES OC(=O)c1ccccc1S
IUPAC Name 2-sulfanylbenzoic acid
Molecular Weight (g/mol) 154.19
Formula Weight 154.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.