missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole, 97%

Product Code. 11443917
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443917 2500 g 2500g
11423917 100 g 100g
11389589 250 g 250g
11433917 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11443917 Supplier Thermo Scientific Alfa Aesar Supplier No. A18350.0E
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 583-39-1 | C7H6N2S | 150.20 g/mol

2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-39-1
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD00466107
InChI Key YHMYGUUIMTVXNW-UHFFFAOYSA-N
PubChem CID 707035
SMILES S=C1NC2=CC=CC=C2N1

Specifications

Melting Point 305°C to 310°C
Density 1.42
Flash Point >250°C (487°F)
Quantity 2500 g
UN Number UN2811
Beilstein 119867
Merck Index 14,1082
Synonym 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
Solubility Information Slightly soluble in water.
IUPAC Name 1,3-dihydrobenzimidazole-2-thione
Formula Weight 150.2
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzimidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.