missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.), 97%, Thermo Scientific™

Product Code. 10088281
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10088281 10 g 10g
10180682 1 g 1g
10727885 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10088281 Supplier Thermo Scientific Maybridge Supplier No. GK00437EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 758-08-7 , 7664-41-7 , 67-56-1
Molecular Formula C2H5NOS
Molecular Weight (g/mol) 91.128
MDL Number MFCD00068159
InChI Key GYXHHICIFZSKKZ-UHFFFAOYSA-N
Synonym 2-mercaptoacetamide, mercaptoacetamide, thioglycolamide, acetamide, 2-mercapto, thioglycolamide crude, usaf ha-3, acetamide, 2-mercapto-6ci,7ci,8ci,9ci, 2-mercaptoacetamide in solution nh3/meoh, thiolacetamide, a-mercaptoacetamide
PubChem CID 12961
IUPAC Name 2-sulfanylacetamide
SMILES C(C(=O)N)S

Specifications

CAS Min % 10.0, 7.0, 75.0
CAS Max % 12.0, 13.0, 83.0
Assay Percent Range 97%
Linear Formula SHCH2C(O)NH2
Quantity 10 g
Formula Weight 91.13
Percent Purity 97%
Chemical Name or Material 2-Mercaptoacetamide (in Methanolic Ammonia Solution 10g/100ml approx.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.