missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Mercapto-6-nitrobenzothiazole, 96%

Product Code. 11423747
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11423747 Supplier Alfa Aesar™ Supplier No. A18211.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4845-58-3
Molecular Formula C7H4N2O2S2
Molecular Weight (g/mol) 212.24
MDL Number MFCD00041850
InChI Key QPOZGXKWWKLJDK-UHFFFAOYSA-N
Synonym 2-mercapto-6-nitrobenzothiazole, 6-nitrobenzo d thiazole-2-thiol, 6-nitro-2-mercaptobenzothiazole, 6-nitrobenzo d thiazole-2 3h-thione, 6-nitrobenzothiazole-2-thiol, 2 3h-benzothiazolethione, 6-nitro, usaf ek-3991, 6-nitro-1,3-benzothiazole-2-thiol, 2-benzothiazolinethione, 6-nitro, 2-benzothiazolethiol, 6-nitro
PubChem CID 947375
IUPAC Name 6-nitro-3H-1,3-benzothiazole-2-thione
SMILES [O-][N+](=O)C1=CC=C2NC(=S)SC2=C1

Specifications

Melting Point 250°C to 254°C
Quantity 1 g
Formula Weight 212.24
Percent Purity 96%
Chemical Name or Material 2-Mercapto-6-nitrobenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.