missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4-phenylthiazole, 98%

Product Code. 11357605
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357605 25 g 25g
11347605 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357605 Supplier Thermo Scientific Alfa Aesar Supplier No. L05829.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 2103-88-0 | C9H7NS2 | 193.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-88-0
Molecular Formula C9H7NS2
Molecular Weight (g/mol) 193.282
MDL Number MFCD00014508
InChI Key CYCKHTAVNBPQDB-UHFFFAOYSA-N
Synonym 4-phenylthiazole-2-thiol, 2-mercapto-4-phenylthiazole, 4-phenyl-1,3-thiazole-2-thiol, 4-phenyl-2-thiazolethiol, 2 3h-thiazolethione, 4-phenyl, 4-phenyl-2-mercaptothiazole, 4-thiazoline-2-thione, 4-phenyl, 2-thiazolethiol, 4-phenyl, 4-phenyl-thiazole-2-thiol, 4-phenylthiazole-2 3h-thione
PubChem CID 3000729
IUPAC Name 4-phenyl-3H-1,3-thiazole-2-thione
SMILES C1=CC=C(C=C1)C2=CSC(=S)N2

Specifications

Melting Point 172°C to 175°C
Odor Odorless
Quantity 25 g
Beilstein 128288
Formula Weight 193.3
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4-phenylthiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.