missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4-methylpyrimidine hydrochloride, 99%

Product Code. 11424524
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424524 100 g 100g
11414524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11424524 Supplier Thermo Scientific Alfa Aesar Supplier No. A13145.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6959-66-6 | C5H7ClN2S | 162.635 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-66-6
Molecular Formula C5H7ClN2S
Molecular Weight (g/mol) 162.635
MDL Number MFCD00012783
InChI Key UQJLPBLXSJWAKG-UHFFFAOYSA-N
Synonym 2-mercapto-4-methylpyrimidine hydrochloride, 4-methylpyrimidine-2-thiol hydrochloride, 4-methylpyrimidine-2 1h-thione hydrochloride, 6-methyl-1,2-dihydropyrimidine-2-thione hydrochloride, 4-methylpyrimidine-2-thiol hcl, 2 1h-pyrimidinethione, 4-methyl-, monohydrochloride, c5h6n2s.clh, 2-mercapto-4-methyl pyrimidine hydrochloride, pubchem21990, acmc-1b3ky
PubChem CID 2723697
IUPAC Name 6-methyl-1H-pyrimidine-2-thione;hydrochloride
SMILES CC1=CC=NC(=S)N1.Cl

Specifications

Melting Point ∼265°C (decomposition)
Quantity 100 g
Beilstein 3691520
Formula Weight 162.64
Percent Purity 99%
Chemical Name or Material 2-Mercapto-4-methylpyrimidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.