missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Mercapto-4,6-dimethylpyrimidine hydrate, 98%

Product Code. 11404923
Change view
Click to view available options
Quantity:
10 g
250 g
Unit Size:
10g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404923 250 g 250g
11484913 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404923 Supplier Alfa Aesar™ Supplier No. A12231.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 22325-27-5
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.204
MDL Number MFCD00006079
InChI Key RAFAYWADRVMWFA-UHFFFAOYSA-N
Synonym 4,6-dimethyl-2-mercaptopyrimidine, 4,6-dimethyl-2 1h-pyrimidinethione, 4,6-dimethylpyrimidine-2-thiol, 4,6-dimethyl-2-thiopyrimidine, 4,6-dimethyl-2-pyrimidinethiol, 2-mercapto-4,6-dimethylpyrimidine, 2 1h-pyrimidinethione, 4,6-dimethyl, 4,6-dimethylpyrimidine-2-thione, 4,6-dimethyl-pyrimidine-2-thiol, 4,6-dimethyl-2-thiolpyrimidine
PubChem CID 673664
IUPAC Name 4,6-dimethyl-1H-pyrimidine-2-thione
SMILES CC1=CC(=NC(=S)N1)C

Specifications

Melting Point 213°C to 216°C
Quantity 250 g
Beilstein 81330
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.