missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-1-methylimidazole, 98%

Product Code. 11454424
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454424 100 g 100g
11444424 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454424 Supplier Thermo Scientific Alfa Aesar Supplier No. A13094.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 60-56-0 | C4H6N2S | 114.17 g/mol

Possesses antioxidant properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00179321
InChI Key PMRYVIKBURPHAH-UHFFFAOYSA-N
Synonym methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol
PubChem CID 1349907
ChEBI CHEBI:50673
IUPAC Name 3-methyl-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Melting Point 144°C to 147°C
Boiling Point 280°C (decomposition)
Odor Stench
Quantity 100 g
Beilstein 108646
Merck Index 14,5971
Solubility Information Freely soluble in water; Soluble in alcohol,chloroform; Sparingly soluble in ether,benzene.
Formula Weight 114.17
Percent Purity 98%
Chemical Name or Material 2-Mercapto-1-methylimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.