missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 2'-MeCCPA
GREENER_CHOICE

Product Code. 18044762
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18044762 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18044762 Supplier Tocris Bioscience™ Supplier No. 2281/10
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Very selective and potent A1 receptor agonist

Potent and highly selective agonist at A1 adenosine receptors (Ki values are 3.3, 9580, 37600 and 1150 nM for human recombinant A1, A2A, A2B and A3 receptors respectively). Acts as a full agonist; inhibits forskolin-stimulated adenylyl cyclase activity in rat cortical membranes with an IC50 value of 13.1 nM.
TRUSTED_SUSTAINABILITY

Specifications

CAS 205171-12-6
Quantity 10 mg
Molecular Formula C16H22ClN5O4
Formula Weight 383.83
Purity >98%
Molecular Weight (g/mol) 383.833
Chemical Name or Material 2'-MeCCPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.