missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric Acid, TRC

Product Code. 17973625
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973625 10 g 10g
17961955 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17973625 Supplier Toronto Research Chemicals Supplier No. TRCK19120010G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-50-7
Molecular Formula C5 H6 O5
Molecular Weight (g/mol) 146.1
Synonym alpha-Ketoglutaric Acid, Pentanedioic acid, 2-oxo- (9CI, ACI), 2-Oxopentanedioic acid (ACI), Glutaric acid, 2-oxo- (8CI), 2-Ketoglutaric acid, 2-Oxo-1,5-pentanedioic acid, 2-Oxoglutaric acid, Aladdin K 105571, NSC 17391, α-keto-Glutaric acid, α-Ketoglutaric acid, α-Oxoglutaric acid, α-Oxopentanedioic acid
IUPAC Name 2-oxopentanedioic acid
SMILES OC(=O)CCC(=O)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)
Formula Weight 146.0215
Percent Purity >95%
Chemical Name or Material alpha-Ketoglutaric acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.