missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric acid, MedChemExpress

Product Code. 16493439
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493439 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493439 Supplier Medchem Express Supplier No. HYW013636
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 2-Ketoglutaric acid (Alpha-Ketoglutaric acid) is an intermediate in the production of ATP or GTP in the Krebs cycle. 2-Ketoglutaric acid also acts as the major carbon skeleton for nitrogen-assimilatory reactions. 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase (IC50=15 mM).

2-Ketoglutaric acid (Alpha-Ketoglutaric acid) is an intermediate in the production of ATP or GTP in the Krebs cycle. 2-Ketoglutaric acid also acts as the major carbon skeleton for nitrogen-assimilatory reactions. 2-Ketoglutaric acid is a reversible inhibitor of tyrosinase (IC50=15 mM).

Specifications

Chemical Name or Material 2-Ketoglutaric acid
CAS 328-50-7
Color White
Purity Grade Notes Research
Molecular Formula C5H6O5
Quantity 1 g
Synonym Alpha-Ketoglutaric acid
Solubility Information DMSO : 250 mg/mL (1711.16 mM; Need ultrasonic) ∣H2O : 50 mg/mL (342.23 mM; Need ultrasonic)
SMILES OC(=O)CCC(=O)C(O)=O
Molecular Weight (g/mol) 146.1
Formula Weight 146.1
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.