missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-L-gulonic Acid, TRC

Product Code. 30393375
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393375 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393375 Supplier Toronto Research Chemicals Supplier No. TRCK19132010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Keto-L-gulonic Acid
CAS 526-98-7
Purity Grade Notes HPLC
Molecular Formula C6H10O7
Quantity 10 g
Synonym L-Xylo-2-Hexulosonic Acid, L-Xylo-Hexulosonic Acid, 2-Keto-L-idonic Acid, 2-Keto-L-gulonic Acid, 2-Keto-L-Gulonic Acid, L-Xylohexulosonic Acid
InChI Formula InChI=1S/C6H10O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-4,7-10H,1H2,(H,12,13)/t2-,3+,4-/m0/s1
SMILES OC[C@H](O)[C@@H](O)[C@H](O)C(=O)C(=O)O
IUPAC Name (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
Molecular Weight (g/mol) 194.14
Formula Weight 194.0427
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.