missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Keto-L-gulonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Keto-L-gulonic Acid |
| CAS | 526-98-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H10O7 |
| Quantity | 250 mg |
| Synonym | L-Xylo-2-Hexulosonic Acid, L-Xylo-Hexulosonic Acid, 2-Keto-L-idonic Acid, 2-Keto-L-gulonic Acid, 2-Keto-L-Gulonic Acid, L-Xylohexulosonic Acid |
| InChI Formula | InChI=1S/C6H10O7/c7-1-2(8)3(9)4(10)5(11)6(12)13/h2-4,7-10H,1H2,(H,12,13)/t2-,3+,4-/m0/s1 |
| SMILES | OC[C@H](O)[C@@H](O)[C@H](O)C(=O)C(=O)O |
| IUPAC Name | (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid |
| Molecular Weight (g/mol) | 194.14 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?