missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-d-gluconic Acid Sodium, TRC

Product Code. 30401104
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401104 25 mg
30399171 200 mg
30408439 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401104 Supplier Toronto Research Chemicals Supplier No. TRCK18855325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6 H10 O7 . x[Na]
Synonym D-Glucosonic Acid Sodium
IUPAC Name (3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid, sodium salt
SMILES [Na].OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H10O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-4,7-10H,1H2,(H,12,13);/t2-,3-,4+;/m1./s1
Percent Purity >95%
Chemical Name or Material 2-Keto-d-gluconic Acid Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.