missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%

Product Code. 15440617
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440617 10 g 10g
15420617 1 g 1g
15430617 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440617 Supplier Thermo Scientific Alfa Aesar Supplier No. J66609.09
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3470-37-9 | C12H20CaO14 | 428.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3470-37-9
Molecular Formula C12H20CaO14
Molecular Weight (g/mol) 428.356
MDL Number MFCD00149104
InChI Key QACANIDDQFGAJU-JQTJLMCZSA-N
Synonym calcium 2-keto-d-gluconate
PubChem CID 131852723
IUPAC Name calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
SMILES C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]

Specifications

Color White
Quantity 10 g
Beilstein 3808734
Optical Rotation −70° (c=2 in Water)
Formula Weight 231.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Keto-D-gluconic acid hemicalcium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.