missing translation for 'onlineSavingsMsg'
Learn More

2-(Isopropylthio)ethanol, 98+%, Thermo Scientific™

Product Code. 11397385
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397385 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11397385 Supplier Thermo Scientific Alfa Aesar Supplier No. L05312.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40811-49-2
Molecular Formula C5H12OS
Molecular Weight (g/mol) 120.21
MDL Number MFCD00014038
InChI Key QGONIKZRWOOHBB-UHFFFAOYSA-N
Synonym 2-isopropylthio ethanol, 2-isopropylsulfanyl ethanol, ethanol, 2-1-methylethyl thio, 2-hydroxyethyl isopropyl sulfide, 2-1-methylethyl thio ethanol, isopropylthioethanol, acmc-1arvo, 2-propan-2-ylthio ethanol, 2-hydroxyethylisopropylsulfide, 2-isopropylsulfanyl ethanol #
PubChem CID 101268
IUPAC Name 2-propan-2-ylsulfanylethanol
SMILES CC(C)SCCO

Specifications

Density 0.979
Boiling Point 191°C to 192°C
Flash Point 94°C (201°F)
Linear Formula (CH3)2CHS(CH2)2OH
Refractive Index 1.478
Quantity 25 g
UN Number UN1760
Beilstein 1697233
Formula Weight 120.21
Percent Purity ≥98%
Chemical Name or Material 2-(Isopropylthio)ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.