missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl isothiocyanate, ≥95%, Thermo Scientific™

Product Code. 10103724
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 36176-31-5
Molecular Formula C10H11NS
Molecular Weight (g/mol) 177.265
MDL Number MFCD00022053
InChI Key QQOOMDSEMQHMIR-UHFFFAOYSA-N
Synonym 2-isopropylphenyl isothiocyanate, 1-isopropyl-2-isothiocyanatobenzene, 1-isothiocyanato-2-propan-2-yl benzene, benzene, 1-isothiocyanato-2-1-methylethyl, 2-methylethyl benzenisothiocyanate, 2-isopropylphenylisothiocyanate, acmc-20ansf, 2-isopropylphenyl-isothiocyanate, 2-iso-propylphenyl isothiocyanate
PubChem CID 142052
IUPAC Name 1-isothiocyanato-2-propan-2-ylbenzene
SMILES CC(C)C1=CC=CC=C1N=C=S

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Linear Formula CH3CH(CH3)C6H4NCS
Quantity 25 g
Formula Weight 177.27
Percent Purity 95%
Chemical Name or Material 2-Isopropylphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'