missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenol, 98+%

Product Code. 11449892
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449892 250 g 250g
11319599 50 g 50g
11439892 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449892 Supplier Thermo Scientific Alfa Aesar Supplier No. A12027.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-69-7 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-69-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00002224
InChI Key CRBJBYGJVIBWIY-UHFFFAOYSA-N
Synonym 2-isopropylphenol, o-hydroxycumene, o-isopropylphenol, o-cumenol, 1-hydroxy-2-isopropylbenzene, 2-propan-2-yl phenol, 2-1-methylethyl phenol, phenol, o-isopropyl, phenol, 2-1-methylethyl, isopropylphenol
PubChem CID 6943
ChEBI CHEBI:38506
IUPAC Name 2-propan-2-ylphenol
SMILES CC(C)C1=CC=CC=C1O

Specifications

Density 1.010
Assay Percent Range 98%
Quantity 250 g
UN Number UN2922
Beilstein 1363322
Formula Weight 136.2
Percent Purity 98%
Chemical Name or Material 2-Isopropylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.