missing translation for 'onlineSavingsMsg'
Learn More

2-isopropylnaphthalene 10000 μg/mL in Hexadecane, Dr. Ehrenstorfer

Product Code. 30324730
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324730 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324730 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010402HD
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-isopropylnaphthalene
CAS 2027-17-0
Analyte or Component Names 2-isopropylnaphthalene
Shipping Condition Room Temperature
Molecular Formula C13 H14
Concentration 10000 μg/mL
Certifications/Compliance ISO 17034
Synonym Naphthalene, 2-(1-methylethyl)-, Naphthalene, 2-isopropyl- (6CI,7CI,8CI), 2-(1-Methylethyl)naphthalene, 2-Isopropylnaphthalene, 6-Isopropylnaphthalene, NSC 166466, β-Isopropylnaphthalene
InChI Formula InChI=1S/C13H14/c1-10(2)12-8-7-11-5-3-4-6-13(11)9-12/h3-10H,1-2H3
SMILES CC(C)c1ccc2ccccc2c1
IUPAC Name 2-propan-2-ylnaphthalene
Solvent or Matrix Hexadecane
Molecular Weight (g/mol) 170.25
Quantity 250 mL
Formula Weight 170.1096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.