missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Isopropylbenzimidazole, 98%

Product Code. 11460153
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460153 25 g 25g
11450153 5 g 5g
11470153 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460153 Supplier Alfa Aesar™ Supplier No. A11356.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5851-43-4
Molecular Formula C10H12N2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00051759
InChI Key RITUGMAIQCZEOG-UHFFFAOYSA-N
Synonym 2-isopropyl-1h-benzo d imidazole, 2-isopropylbenzimidazole, 2-isopropyl-1h-benzimidazole, benzimidazole, 2-isopropyl, 1h-benzimidazole, 2-1-methylethyl, 2-propan-2-yl-1h-1,3-benzodiazole, 2-isopropyl-1h-1,3-benzodiazole, 1h-benzimidazole, 2-1-methylethyl-9ci, 2-methylethyl benzimidazole, acmc-1avkb
PubChem CID 22121
IUPAC Name 2-propan-2-yl-1H-benzimidazole
SMILES CC(C)C1=NC2=CC=CC=C2N1

Specifications

Melting Point 233°C to 237°C
Quantity 25 g
Beilstein 3535
Formula Weight 160.22
Percent Purity 98%
Chemical Name or Material 2-Isopropylbenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.