missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylaniline, 97%, ACROS Organics™

Brand:  Acros Organics 259391000

Code : 29

Additional Details : CAS Number : 643-28-7 Weight : 0.14550kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10333812

  • £26.90 / 100mL


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Isopropylaniline
96.0
97%
C9H13N
135.21
o-Aminocumene
643-28-7
100.0
97%
(CH3)2CHC6H4NH2
MFCD00007720

Specifications

2-Isopropylaniline
112.0° to 113.0°C (18.0 mmHg)
100mL
96.0
0.9550g/mL
Authentic
97%
(CH3)2CHC6H4NH2
MFCD00007720
o-Aminocumene
1.5470 to 1.5500 (20°C, 589nm)
95°C
Clear pale yellow to orange or brown
643-28-7
100.0
Liquid
97%
C9H13N
135.21
Glass bottle
0.955
Product may darken on storage
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Isopropylaniline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS