missing translation for 'onlineSavingsMsg'
Learn More

2-(Isopropylamino)acetophenone hydrochloride, 98%, Thermo Scientific™

Product Code. 11474780
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474780 250 mg 250mg
11484780 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474780 Supplier Thermo Scientific Alfa Aesar Supplier No. H50799.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38506-97-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD18785640
InChI Key IHVMTNLVDVWAAA-UHFFFAOYSA-N
Synonym 2-isopropylamino-1-phenylethanone, 2-isopropylamino acetophenone, phenyl isopropylaminomethyl ketone, 2-methylethyl amino-1-phenylethan-1-one, 1-phenyl-2-propan-2-yl amino ethan-1-one, 2-isopropylamino acetophenone hydrochloride
PubChem CID 12861561
IUPAC Name 1-phenyl-2-(propan-2-ylamino)ethanone
SMILES CC(C)NCC(=O)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 213.71
Percent Purity 98%
Chemical Name or Material 2-(Isopropylamino)acetophenone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.