missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylamino-2'-methoxyacetophenone, Thermo Scientific™

Product Code. 11444660
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444660 1 g 1g
11434660 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444660 Supplier Thermo Scientific Alfa Aesar Supplier No. H50734.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-89-9
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD08064237
InChI Key FGSJNHHRBVOFTM-UHFFFAOYSA-N
Synonym 2-isopropylamino-2'-methoxyacetophenone, 2-isopropylamino-1-2-methoxyphenyl ethanone, 2-isopropylamino-1-2-methoxy-phenyl-ethanone, 1-2-methoxyphenyl-2-methylethyl amino ethan-1-one
PubChem CID 24208808
IUPAC Name 1-(2-methoxyphenyl)-2-(propan-2-ylamino)ethanone
SMILES CC(C)NCC(=O)C1=CC=CC=C1OC

Specifications

Quantity 1 g
Formula Weight 207.27
Chemical Name or Material 2-Isopropylamino-2'-methoxyacetophenone
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.