missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Isopropylamino-1-phenylethanol

Product Code. 11484770
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11484770

Brand: Alfa Aesar™ H50793.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4164-21-0
Molecular Formula C11H17NO
Molecular Weight (g/mol) 179.26
MDL Number MFCD01732724
InChI Key YANYGOBZUASRHR-UHFFFAOYNA-N
Synonym 2-isopropylamino-1-phenylethanol, 1-phenyl-2-propan-2-ylamino ethanol, 1-phenyl-2-isopropylaminoethanol, alpha-isopropylaminomethyl benzyl alcohol, alpha-1-methylethyl amino methyl benzenemethanol, 1-phenyl-1-oxy-2-isopropylamino-aethan german, 1-phenyl-2-propan-2-yl-amino ethan-1-ol, benzyl alcohol, alpha-isopropylaminomethyl, 2-methylethyl amino-1-phenylethan-1-ol
PubChem CID 20119
IUPAC Name 1-phenyl-2-(propan-2-ylamino)ethanol
SMILES CC(C)NCC(O)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 179.26
Chemical Name or Material 2-Isopropylamino-1-phenylethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.