missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 2-Isopropylamino-1-phenylethanol
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Specifications
Specifications
| Quantity | 250 mg |
| Formula Weight | 179.26 |
| Chemical Name or Material | 2-Isopropylamino-1-phenylethanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction