missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-d7 Thioxanthone, TRC

Product Code. 30399603
Change view
Click to view available options
Quantity:
1 mg
10 mg
Product Code. Quantity
30399603 1 mg
30394922 10 mg
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30399603

Brand: Toronto Research Chemicals TRCI8735021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173019-24-3
InChI Formula InChI=1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3/i1D3,2D3,10D
Molecular Formula C16H7D7OS
Molecular Weight (g/mol) 261.39
SMILES O=C1C2=C(C=CC(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=C2)SC3=CC=CC=C31
Synonym 2-(1-Methylethyl-d7)-9H-thioxanthen-9-one, 2-(Isopropyl-d7)-9H-thioxanthen-2-one, 2-(Isopropyl-d7)thioxanthen-9-one, ITX-d7, Photocure ITX-P-d7, Quantacure 597-d7, Quantacure 659-d7, Quantacure ITX-d7, Speedcure ITX-d7
Chemical Name or Material 2-Isopropyl-d7 Thioxanthone

Specifications

Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.