missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-1,3-dioxane-5,5-dicarboxylic Acid 5,5-Diethyl Ester, TRC

Product Code. 30394680
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394680 5 g -
30401303 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394680 Supplier Toronto Research Chemicals Supplier No. TRCI8244255G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35113-48-5
Molecular Formula C13H22O6
Molecular Weight (g/mol) 274.31
Synonym 2-(1-Methylethyl)-1,3-dioxane-5,5-dicarboxylic Acid Diethyl Ester, 2-Isopropyl-5,5-dicarbethoxy-1,3-dioxane
IUPAC Name diethyl 2-propan-2-yl-1,3-dioxane-5,5-dicarboxylate
SMILES CCOC(=O)C1(COC(OC1)C(C)C)C(=O)OCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H22O6/c1-5-16-11(14)13(12(15)17-6-2)7-18-10(9(3)4)19-8-13/h9-10H,5-8H2,1-4H3
Formula Weight 274.1416
Chemical Name or Material 2-Isopropyl-1,3-dioxane-5,5-dicarboxylic Acid 5,5-Diethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.