missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxypyridine-5-boronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15461679
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
870521-30-5
C8H12BNO3
181.00
MFCD07368870
SGEOBUJTURUUJE-UHFFFAOYSA-N
6-isopropoxypyridin-3-yl boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxy-3-pyridyl boronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isopropoxy-5-pyridineboronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, 6-isoproxypyridine-3-boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
44754864
(6-propan-2-yloxypyridin-3-yl)boronic acid
CC(C)OC1=NC=C(C=C1)B(O)O
This item is not returnable. View return policy
870521-30-5
C8H12BNO3
181.00
MFCD07368870
SGEOBUJTURUUJE-UHFFFAOYSA-N
6-isopropoxypyridin-3-yl boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxy-3-pyridyl boronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isopropoxy-5-pyridineboronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, 6-isoproxypyridine-3-boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
44754864
(6-propan-2-yloxypyridin-3-yl)boronic acid
CC(C)OC1=NC=C(C=C1)B(O)O

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 181
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Isopropoxypyridine-5-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'