missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenol, 97%

Product Code. 11361877
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361877 5 g 5g
11371877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361877 Supplier Thermo Scientific Alfa Aesar Supplier No. L07360.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4812-20-8 | C9H12O2 | 152.193 g/mol

2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyphenol is a metabolite of propoxur. It was used as a target chemical in developing a method to measure the urinary phenolic metabolites

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4812-20-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00002184
InChI Key ZNCUUYCDKVNVJH-UHFFFAOYSA-N
Synonym 2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a
PubChem CID 20949
ChEBI CHEBI:38547
IUPAC Name 2-propan-2-yloxyphenol
SMILES CC(C)OC1=CC=CC=C1O

Specifications

Density 1.032
Boiling Point 100°C to 102°C (11 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.515
Quantity 5 g
Beilstein 1937063
Solubility Information Soluble in water.
Formula Weight 152.19
Percent Purity 97%
Chemical Name or Material 2-Isopropoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.