missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyphenol, 97%, Thermo Scientific™

Product Code. 10185120 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10185120 25 g Glass bottle 25g
10323722 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10185120 Supplier Thermo Scientific Acros Supplier No. 209220250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4812-20-8
MDL Number MFCD00002184
InChI Key ZNCUUYCDKVNVJH-UHFFFAOYSA-N
Synonym 2-isopropoxyphenol, o-isopropoxyphenol, phenol, 2-1-methylethoxy, 2-1-methylethoxy phenol, 2-propan-2-yloxy phenol, unii-x51p4ue6x7, phenol, 2-isopropoxy, 2-propan-2-yl-oxy phenol, isopropyl catechol, caswell no. 508a
PubChem CID 20949
ChEBI CHEBI:38547
IUPAC Name 2-propan-2-yloxyphenol
SMILES CC(C)OC1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown-Red to Yellow
Density 1.0300g/mL
Boiling Point 100.0°C to 102.0°C (11.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O2
Linear Formula (CH3)2CHOC6H4OH
Refractive Index 1.5140 to 1.5160
Quantity 25 g
Specific Gravity 1.03
Molecular Weight (g/mol) 152.19
Formula Weight 152.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Isopropoxyphenol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.