missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethanol, 99%

Product Code. 11347986
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347986 2500 mL 2.5 L
11327986 100 mL 100mL
11337986 1000 mL 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347986 Supplier Thermo Scientific Alfa Aesar Supplier No. L13241.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-59-1 | C5H12O2 | 104.15 g/mol

2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-59-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002866
InChI Key HCGFUIQPSOCUHI-UHFFFAOYSA-N
Synonym 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
PubChem CID 7996
IUPAC Name 2-propan-2-yloxyethanol
SMILES CC(C)OCCO

Specifications

Melting Point -60°C
Density 0.903
Boiling Point 143°C
Flash Point 45°C (113°F)
Linear Formula (CH3)2CHO(CH2)2OH
Refractive Index 1.41
Quantity 2500 mL
UN Number UN1992
Beilstein 1732184
Solubility Information It is soluble in water.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material 2-Isopropoxyethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.