missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 94%

Product Code. 11344016
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344016 5 g 5g
11334016 1 g 1g
11354016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11344016 Supplier Thermo Scientific Alfa Aesar Supplier No. L17278.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 61676-62-8 | C9H19BO3 | 186.058 g/mol

Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61676-62-8
Molecular Formula C9H19BO3
Molecular Weight (g/mol) 186.058
MDL Number MFCD00192241
InChI Key MRWWWZLJWNIEEJ-UHFFFAOYSA-N
Synonym 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane
PubChem CID 5017741
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)OC(C)C

Specifications

Density 0.912
Boiling Point 73°C (15 mmHg)
Flash Point 42°C (107°F)
Refractive Index 1.409
Quantity 5 g
UN Number UN1993
Beilstein 1906370
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.