missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Product Code. 10331614 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10331614 5 g Glass bottle 5g
10437012 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10331614 Supplier Thermo Scientific Acros Supplier No. 359490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 61676-62-8
Molecular Formula C9H19BO3
Molecular Weight (g/mol) 186.06
MDL Number MFCD00192241
InChI Key MRWWWZLJWNIEEJ-UHFFFAOYSA-N
Synonym 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane
PubChem CID 5017741
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)OC(C)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9120g/mL
Boiling Point 73°C (15.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4080 to 1.41
Quantity 5 g
Specific Gravity 0.912
Formula Weight 186.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.