missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%, Thermo Scientific™

Catalog Number 10437012
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
61676-62-8
C9H19BO3
186.06
MFCD00192241
MRWWWZLJWNIEEJ-UHFFFAOYSA-N
2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane
5017741
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)OC(C)C
This item is not returnable. View return policy
61676-62-8
C9H19BO3
186.06
MFCD00192241
MRWWWZLJWNIEEJ-UHFFFAOYSA-N
2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane
5017741
4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)OC(C)C
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9120g/mL
Boiling Point 73°C (15.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4080 to 1.41
Quantity 1 g
Specific Gravity 0.912
Formula Weight 186.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'