missing translation for 'onlineSavingsMsg'
Learn More

2-Isonitrosoacetophenone, TRC

Product Code. 30408511
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408511 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408511 Supplier Toronto Research Chemicals Supplier No. TRCI8218601G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Isonitrosoacetophenone
CAS 532-54-7
Molecular Formula C8H7NO2
Quantity 1 g
Synonym Phenylglyoxal 2-Oxime, Phenylglyoxal Aldoxime, 2-(Hydroxyimino)acetophenone, Benzoylformaldoxime, Isonitrosoacetophenone, NSC 1330, NSC 25388, Phenylglyoxal Monoxime, RA 54, alpha-(Hydroximino)acetophenone, alpha-(Hydroxyimino)acetophenone, (E)-2-Oxo-2-phenylacetaldehyde Oxime
InChI Formula InChI=1S/C8H7NO2/c10-8(6-9-11)7-4-2-1-3-5-7/h1-6,11H
SMILES ON=CC(=O)c1ccccc1
IUPAC Name 2-hydroxyimino-1-phenylethanone
Molecular Weight (g/mol) 149.15
Formula Weight 149.0477
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.