missing translation for 'onlineSavingsMsg'
Learn More

2-Isoniazid, TRC

Product Code. 30395128
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395128 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395128 Supplier Toronto Research Chemicals Supplier No. TRCI8214551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Isoniazid
CAS 1452-63-7
Molecular Formula C6 H7 N3 O
Quantity 1 g
Synonym 2-Pyridinecarboxylic Acid Hydrazide, Picolinic acid, hydrazide (6CI,7CI,8CI), 2-(Hydrazinocarbonyl)pyridine, 2-Pyridinecarboxylic hydrazide, 2-Pyridylcarboxylic acid hydrazide, INHd 2, NSC 81875, Picolinic hydrazide, Picolinoyl hydrazide, Picolinoylhydrazine, Pyridine-2-carbohydrazide, WS 104, α-Picolinohydrazide, 2-Isoniazid
InChI Formula InChI=1S/C6H7N3O/c7-9-6(10)5-3-1-2-4-8-5/h1-4H,7H2,(H,9,10)
SMILES NNC(=O)c1ccccn1
IUPAC Name pyridine-2-carbohydrazide
Molecular Weight (g/mol) 137.14
Formula Weight 137.0589
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.