missing translation for 'onlineSavingsMsg'
Learn More

2-Isocyanatobenzonitrile, 97%, Thermo Scientific™

Product Code. 10066664
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10066664 5 g 5g
10548253 250 mg 250mg
10502632 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10066664 Supplier Thermo Scientific Maybridge Supplier No. BTB13667DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 42066-86-4
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
InChI Key FQLNVWAFXQUXGW-UHFFFAOYSA-N
PubChem CID 2735873
IUPAC Name 2-isocyanatobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)N=C=O

Specifications

Assay Percent Range 95%
Linear Formula CNC6H4NCO
Quantity 5 g
Synonym 2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b
Formula Weight 144.13
Percent Purity 95%
Chemical Name or Material 2-Isocyanatobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.