missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutylpyridine, 99%, Thermo Scientific Chemicals

Product Code. 11446066
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11446066

missing translation for 'mfr': Thermo Scientific Alfa Aesar B22349.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6304-24-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD02262142
InChI Key BBVSPSDWPYWMOR-UHFFFAOYSA-N
Synonym 2-isobutylpyridine, 2-2-methylpropyl pyridine, 2-isobutyl pyridine, pyridine, 2-2-methylpropyl, unii-gs3k23h1q8, pyridine, 2-isobutyl, fema no. 3370, 2-isobutyl-pyridin, 2-isobutyl-pyridine, acmc-1b4hh
PubChem CID 61385
IUPAC Name 2-(2-methylpropyl)pyridine
SMILES CC(C)CC1=CC=CC=N1

Specifications

Density 0.899
Quantity 5 g
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material 2-Isobutylpyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.