missing translation for 'onlineSavingsMsg'
Learn More

2-Iodotoluene, TRC

Product Code. 30403099
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403099 1 g -
30401505 5 g 5g
30392604 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403099 Supplier Toronto Research Chemicals Supplier No. TRCI7243001G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-37-2
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.03
Synonym 1-Iodo-2-methylbenzene, 2-Methyl-1-iodobenzene, 2-Methyliodobenzene, 2-Methylphenyl Iodide, 2-Tolyl Iodide, NSC 3774, o-Iodotoluene, o-Methyliodobenzene, o-Methylphenyl Iodide, o-Tolyl Iodide
IUPAC Name 1-iodo-2-methylbenzene
SMILES Cc1ccccc1I

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H7I/c1-6-4-2-3-5-7(6)8/h2-5H,1H3
Formula Weight 217.9592
Chemical Name or Material 2-Iodotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.