missing translation for 'onlineSavingsMsg'
Learn More

2-Iodotoluene, 98%

Product Code. 11495337
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495337 25 g 25g
11405347 100 g 100g
11415347 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495337 Supplier Thermo Scientific Alfa Aesar Supplier No. B22697.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 615-37-2 | C7H7I | 218.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-37-2
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001042
InChI Key RINOYHWVBUKAQE-UHFFFAOYSA-N
Synonym 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07
PubChem CID 5128
ChEBI CHEBI:39813
IUPAC Name 1-iodo-2-methylbenzene
SMILES CC1=CC=CC=C1I

Specifications

Density 1.713
Boiling Point 210°C to 212°C
Flash Point 90°C (194°F)
Refractive Index 1.609
Quantity 25 g
Beilstein 1634294
Sensitivity Light sensitive
Formula Weight 218.04
Percent Purity 98%
Chemical Name or Material 2-Iodotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.