missing translation for 'onlineSavingsMsg'
Learn More

2-Iodosobenzoic acid, 97%

Product Code. p-7046659
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11348346

Brand: Thermo Scientific Alfa Aesar L15606.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304-91-6 | C7H5IO3 | 264.018 g/mol

1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Organosulfonyloxy-1, 2-benziodoxol-3-(1H)-ones (3, 4, 5) can be prepared in one step from 2-iodosobenzoic acid and the corresponding sulfonic acids. 2-carboxydiphenyliodonium salts was prepared by the acid-catalyzed condensation of 2-iodosobenzoic acid with benzene, mesitylene, and cyclohexylbenzene.

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-91-6
Molecular Formula C7H5IO3
Molecular Weight (g/mol) 264.018
MDL Number MFCD00002401
InChI Key IFPHDUVGLXEIOQ-UHFFFAOYSA-N
Synonym 2-iodosobenzoic acid, o-iodosobenzoic acid, o-iodosylbenzoic acid, o-iodosobenzoate, benzoic acid, 2-iodosyl, benzoic acid, o-iodoso, 2-iodosyl benzoate, benzoic acid, iodosyl, ortho-iodosylbenzoic acid, 3h-1,2-benziodoxol-1-ium, 3-oxo-, hydroxide
PubChem CID 67537
ChEBI CHEBI:52698
IUPAC Name 2-iodosylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)I=O

Specifications

Melting Point ∽230°C (decomposition)
Quantity 1 g
Beilstein 1939973
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 264.02
Percent Purity 97%
Chemical Name or Material 2-Iodosobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.