missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyridine, 95%

Product Code. 15459538
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459538 25 g 25g
15449538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459538 Supplier Thermo Scientific Alfa Aesar Supplier No. H63062.14
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5029-67-4 | C5H4IN | 205.00 g/mol

2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopyridine is a reagent used in the preparation of human NAD+-dependent 15-hydroxyprostaglandin dehydrogenase inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5029-67-4
Molecular Formula C5H4IN
Molecular Weight (g/mol) 205.00
MDL Number MFCD00464928
InChI Key CCZWSTFVHJPCEM-UHFFFAOYSA-N
Synonym 2-iodo-pyridine, pyridine, iodo, iodopyridine, 2-iodo pyridine, 2-iodopyridine, 2-pyridyl iodide, 2-pyridyl iodide;, pyridine, 2-iodo, pubchem4074, acmc-1avou
PubChem CID 221126
IUPAC Name 2-iodopyridine
SMILES IC1=CC=CC=N1

Specifications

Melting Point 52°C (decomposition)
Color Yellow
Density 1.928 g/mL
Refractive Index 1.632
Quantity 25 g
Beilstein 105790
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 205
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Iodopyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.