missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyridine-4-carboxylic acid, 95%, Thermo Scientific Chemicals

Product Code. 15494017
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15494017

Brand: Thermo Scientific Alfa Aesar H31002.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58481-10-0
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD06800803
InChI Key FALCXGCLOUKBSP-UHFFFAOYSA-N
Synonym 2-iodoisonicotinic acid, 2-iodo-isonicotinic acid, 2-iodo-4-pyridinecarboxylic acid, 4-pyridinecarboxylic acid, 2-iodo, 2-iodopyridine-4-carboxylicacid, pubchem16692, 4-pyridinecarboxylicacid, 2-iodo, 2-iodopyridine-4-carboxylic acid
PubChem CID 14530245
IUPAC Name 2-iodopyridine-4-carboxylic acid
SMILES C1=CN=C(C=C1C(=O)O)I

Specifications

Melting Point ∽210°C (decomposition)
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 249.01
Percent Purity 95%
Chemical Name or Material 2-Iodopyridine-4-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.