missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stabilized

Product Code. 10124180 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124180 500 mL Glass bottle 500mL
10011880 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124180 Supplier Thermo Scientific Supplier No. 164965000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-30-9 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 75-30-9 , 7440-50-8
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Especificaciones

CAS Min % 98.0
CAS Max % 100.0
Melting Point -90.0°C
Color Colorless to Yellow
Density 1.7000g/mL
Boiling Point 90.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHI
Refractive Index 1.4972 to 1.4992
Quantity 500 mL
Beilstein 01, 114
Merck Index 15, 5260
Specific Gravity 1.7
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with chloroform,ehter,alcohol and,benzene
Stabilizer copper
Formula Weight 169.99
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, 98+%
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.