missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, Extra Pure, SLR, Fisher Chemical

Product Code. 10254340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic Coated Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10254340 100 mL Plastic Coated Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10254340 Supplier Fisher Chemical Supplier No. I/0831/PB05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 75-30-9
Melting Point -90°C
Color Yellow
Boiling Point 90°C
Packaging Plastic Coated Glass bottle
Molecular Formula C3H7I
MDL Number 1071
Quantity 100 mL
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
SMILES CC(C)I
IUPAC Name 2-iodopropane
Molecular Weight (g/mol) 169.993
PubChem CID 6362
Formula Weight 169.99g/mol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Iodopropane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.