missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetonitrile, 96%

Product Code. 11323366
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323366 5 g 5g
11313366 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323366 Supplier Thermo Scientific Alfa Aesar Supplier No. L12767.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40400-15-5 | C8H6IN | 243.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40400-15-5
Molecular Formula C8H6IN
Molecular Weight (g/mol) 243.047
MDL Number MFCD00040888
InChI Key FPSGTRJUQLYLHE-UHFFFAOYSA-N
Synonym 2-iodophenylacetonitrile, 2-2-iodophenyl acetonitrile, 2-iodobenzyl cyanide, o-iodophenylacetonitrile, benzeneacetonitrile, 2-iodo, o-iodobenzyl cyanide, acmc-20a7xu, 2-iodo-phenylacetonitrile, 2-iodo-phenyl-acetonitrile, 2-2-iodophenyl ethanenitrile
PubChem CID 4402282
IUPAC Name 2-(2-iodophenyl)acetonitrile
SMILES C1=CC=C(C(=C1)CC#N)I

Specifications

Density 1.75
Boiling Point 139°C to 140°C (13 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.617
Quantity 5 g
UN Number UN3276
Beilstein 2083106
Sensitivity Light sensitive
Formula Weight 243.05
Percent Purity 96%
Chemical Name or Material 2-Iodophenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.