missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11348146
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11348146 5 g 5g
11338146 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348146 Supplier Thermo Scientific Alfa Aesar Supplier No. L15116.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18698-96-9 | Molecular Formula: C8H7IO2 | Molecular Weight: 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18698-96-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00046546
InChI Key IUHXGZHKSYYDIL-UHFFFAOYSA-N
Synonym 2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
PubChem CID 2780090
IUPAC Name 2-(2-iodophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)I

Specifications

Melting Point 115°C to 117°C
Quantity 5 g
Beilstein 2086800
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity ≥98%
Chemical Name or Material 2-Iodophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.