missing translation for 'onlineSavingsMsg'
Learn More

(2-Iodoethyl)benzene, 97%, stabilized with copper, ACROS Organics™

Brand:  Acros Organics 305250050

Code : 29

Additional Details : CAS Number : 17376-04-4 Weight : 0.06000kg

Packaging Glass bottle
Quantity 5mL

 View more versions of this product

Product Code. 10063650

  • £39.28 / 5mL

Estimated Shipment: 16-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

17376-04-4
232.07
KVTHPKXDLVYNCH-UHFFFAOYSA-N
28503
C1=CC=C(C=C1)CCI
C8H9I
MFCD00039408
2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene
2-iodoethylbenzene

Specifications

(2-Iodoethyl)benzene
110°C
Glass bottle
1.6
Brown-Red
97%
96.0
96% min. (GC)
C6H5CH2CH2I
2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene
KVTHPKXDLVYNCH-UHFFFAOYSA-N
2-iodoethylbenzene
28503
Liquid
1.6000g/mL
Authentic
1.6000 to 1.6020
122.0°C to 123.0°C (13.0 mmHg)
5mL
17376-04-4
100.0
C8H9I
MFCD00039408
Solubility in water: insoluble in water
C1=CC=C(C=C1)CCI
232.07
232.07
97%
Videos
SDS Documents
Certificates